Thermo Fisher Scientific
How to fix Triplus RSH issue on the internal standard post liquid injection option
Issue
  • In the Liquid, internal standard post, the mode doesn’t work correctly.
  • In many cases, the syringe didn’t pickup correctly the sample during the last step of this injection mode.
Environment
  • Triplus RSH
  • Triplus 100
Resolution
The actual solution until the release of the Triplus RSH 2nd Generation, is to create and load the following script:
clipboard_e6efe7f26d959a8260ad8a1fd6dcd6984.png
Cause
Wrong instructions in the Firmware script.