Method Setup Page and Parameter Check Boxes for Trace GC 1300 / 1600
Issue
When the radio button for a parameter is left with a blue highlight the parameter value is extracted for the value currently on the Trace 1300 series.
Environment
- Trace 1300 Series
- Chromeleon 7.2
Resolution
Do not use the blue highlighted check box. Either leave the box checked/activated or unchecked/deactivated and the instrument will use the parameters within the Chromeleon method.
Attachment(s)
File | Last Modified |
---|---|
Method Setup Page and Parameter Check Boxes - image 1.png | January 10, 2022 |