Thermo Fisher Scientific
Part number for the Cooling Fan inside the electronic module
The part number for the electronic module cooling fan for the TRACE 1300 and TRACE 1310 is 23043713.
Issue
What is the part number for the cooling fan inside the electronic module?
Environment
  • Trace 1300
  • Trace 1310
Resolution
The part number for the electronic module cooling fan is 23043713.
  • Was this article helpful?