Thermo Fisher Scientific
PTV Inlet Body Part Number
Inform the customer that the PTV body for the Trace 1300, which can be replaced in cases of heater or thermocouple malfunction, has the part number 36812739. Note that the part description is incorrectly listed as “Head Support Assembly."
Issue
  • A customer asked for this part to have a glass coating process performed on the PTV inlet.
  • The PTV body can also be replaced in cases where the heater or the thermocouple have malfunctioned.
Environment
Trace 1300
Resolution
The P/N for PTV body is 36812739.
NOTE: The description of the part is incorrect and states the part description as “Head Support Assembly."
clipboard_e4fd933f23005527855e3113f9dd9ca0a.png
Figure 1: Part and part number