Dionex™ Trap Column/Suppressor Cleanup Kit
Issue
Use the Thermo Scientific™ Dionex™ Trap Column/Suppressor Cleanup Kit (PN 059659) to deliver reagent solutions to regenerate and cleanup our trap columns and suppressors. This stand-alone pneumatically driven reagent delivery system eliminates the need for using the analytical pump for delivery of concentrated acids, bases or other reagents that can contaminate the IC system. The kit consists of a 1L plastic reservoir, a gas pressure regulator, a two-way liquid shut-off valve, and tubing and connection fittings. The kit is pre-assembled and easy to install and operate.
Environment
Dionex™ Trap Column/Suppressor Cleanup Kit
Resolution
Dionex™ Trap Column/Suppressor Cleanup Kit
Attachment(s)
File | Last Modified |
---|---|
Man-031835-Installation-Trap-Column-Suppressor-Clean-Up-Kit-Man031835-EN.pdf | July 11, 2023 |