Thermo Fisher Scientific
Error Code 83
Issue
  • Error Code 83: Column temperature below normal range reported (0.05°C)
  • Temperature Sensor showed short after Installation from time to time 
Environment
  • Ion Chromatography
  • ICS-6000
  • ICS-5000
  • DC Module 
Resolution
  • Contact Local Service Support Non-Customer / Internal
  • Back Plane TEC PCB and or Temperature Sensor Cable to be replaced; then Sensor recalibrated
Procedure is the same for the ICS3000/ICS5000/ ICS5000 Plus; however error codes reported may be different – symptoms and fix are the same. 
Cause
  • Possible Failure of TEC PCB
  • Damage to temperature sensor cable