Thermo Fisher Scientific
Needle on the AS-AP sampler is breaking
Issue
Needle on the AS-AP sampler is breaking 
Environment
AS-AP Autosampler
Resolution
An Alignment for the sampler tray and the injection port is needed and sampler needle must be changed. 
Analysis
The use of unknown vials size and different suppliers could cause such problems. The habit of moving the trays or needle arm manually on AS-AP sampler, while its running, will effect on the alignment of the needle and will lead to misalignment and needle breaking.
Cause
Causes of this issue includes:
  • Manual movement of sampler trays and the needle. Always use the commands from the software in Chromeleon to move the tray, as the alignment values of the needle might be compromised if moved manually. 
  • Use of unkonwn vial size. Always use certified Thermofisher vials as recommended in the sampler user manuals.
  • Was this article helpful?