Command rejected
Issue
Command rejected – device is busy.
Environment
- Vanquish Core
- VC-D11
- VC-D12
- Vanquish Flex
- VF-D11
Resolution
- Wait for 5 minutes before you try the previous command again.
- If the problem persists, turn off the module.
- Wait for 5 seconds and turn on the module again.