Clearing Rack Information
Issue
Clearing rack information
Environment
- Gallery/Gallery Plus/Beermaster Analyzer 7.0
- Gallery/Gallery Plus/Beermaster Analyzer 6.0
Resolution
To clear the information from all positions from a rack:
NOTE: The rack has to be outside of the analyzer when the rack information is handled.
- Select F3 > REAGENT RACKS.
- Select the rack from the racks field.
- Select the rack position.
- Click 'clear position' to remove the rack position information.