Power Down to Complete Shut Off
Issue
How do I power down to complete shut off?
Environment
- Heracell VIOS 160i
- Heracell VIOS 250i
Resolution
For more information, refer to the Heracell VIOS 160i/250i User Manual.
- Remove the culture containers and all accessories from the workspace.
- Procure a collecting vessel of sufficient capacity.
- Hold open end of hose into collecting vessel and lock drain hose valve safely into receptacle. The water reservoir starts draining.
Image 1: Fill and drain valve of water reservoir
-
Allow the water reservoir to drain completely into the collecting vessel.
-
Remove drain hose valve again.
-
Start the Steri-Run decontamination routine.
-
Turn the device off using the power switch when the steri-run decontamination routine has been completed.
-
Unplug the power connector and protect it against accidental reconnection.
-
Close shut-off valves of the CO2 / O2 / N2 supply system.
-
Disconnect the gas pressure hoses from the sleeve at the rear of the device.
-
Until the device is shut down, the workspace must be continuously ventilated.
-
Leave the glass door and the outer door open and secure them in this state.