User Interface, Display failure
Issue
Unit experiencing user interface (i.e., display) failure
Environment
TSX Ultra Low Temperature (ULT) Series:
Thermo Scientific – TSX40086A
Thermo Scientific – TSX40086D
Thermo Scientific – TSX40086V
Thermo Scientific – TSX40086G
Thermo Scientific – TSX50086A
Thermo Scientific – TSX50086D
Thermo Scientific – TSX50086V
Thermo Scientific – TSX50086G
Thermo Scientific – TSX60086A
Thermo Scientific – TSX60086D
Thermo Scientific – TSX60086V
Thermo Scientific – TSX60086G
Thermo Scientific – TSX70086A
Thermo Scientific – TSX70086D
Thermo Scientific – TSX70086V
Thermo Scientific – TSX70086G
Resolution
Possible Causes and Solutions:
- Cause: Breaker switch off
Solution: Check circuit breaker and reset to on position. Always use a dedicated, properly grounded circuit. - Cause: User interface not powered on
Solution: Push power button on user interface and hold for at least 1 second.