Unit warms up
Issue
Unit warms up
Environment
| TSX Laboratory Refrigerators | TSX -30° Lab Freezers | TSX -30° Plasma Freezers | TSX -20° Laboratory and Enzyme Freezers | Models | Energy Star Model |
| TSX1205G | TSX1230F | TSX1230L | TSX2320F | V/Y/Z | A/D |
| TSX1205S | TSX2330F | TSX2330L | TSX3020F | V/Y/Z | A/D |
| TSX1205P | TSX3030F | TSX3030L | TSX2320E | V/Y/Z | A/D |
| TSX2305G | TSX5030F | TSX5030L | TSX3020E | V/Y/Z | A/D |
| TSX2305S | V/Y/Z | A/D | |||
| TSX2305P | V/Y/Z | A/D | |||
| TSX2305C | V/Y/Z | A/D | |||
| TSX3005G | V/Y/Z | A/D | |||
| TSX3005S | V/Y/Z | A/D | |||
| TSX3005P | V/Y/Z | A/D | |||
| TSX3005C | V/Y/Z | A/D | |||
| TSX4505G | V/Y/Z | A/D | |||
| TSX4505C | V/Y/Z | A/D | |||
| TSX5005G | V/Y/Z | A/D | |||
| TSX5005S | V/Y/Z | A/D | |||
| TSX5005P | V/Y/Z | A/D | |||
| TSX5005C | V/Y/Z | A/D |
Resolution
Check the following:
1. Door is open: Make sure the door is completely closed.
2. Door seal: Check the door seal. To check the door seal, complete the following steps:
o Open the door. o Insert a strip of paper (a couple of inches wide) between the door gasket and the cabinet flange and close the door.
o Slowly pull the paper strip from the outside. You should feel some resistance.
o Repeat this test at 4-inch intervals around the door. If the door does not seal properly, replace the gasket.
3. Warm product recently loaded in unit: Allow ample time to recover from loading warm product.
4. Power supply: Check for proper voltage to the unit. If there is no voltage to the unit, call an electrician.
5. If setpoints need to be adjusted, follow these steps to adjust the setpoint:
o You can enter the programming mode by pressing the Scan button (SCAN) and holding for 5 seconds. The display will then flash "Prg" to indicate that you have entered the programming mode.
o Use (+) and (-) buttons to modify the values and press the scan (SCAN )button to save the value and go to the next screen.
| Screen Button | Function | Summary | |
| 1 | Initial Screen | Unit Set point | The temperature that the unit is set to run |
| 2 | Press Scan | Cold Alarm Temperature | The temperature that will actuate the cold alarm (if active) |
| 3 | Press Scan | Warn Alarm Temperature | The temperature that will actuate the warm alarm (if active) |
| 4 | Press Scan | Exit Program Mode | Returns to normal operating screen |
Note: If the unit is left idle for a period of 30 seconds during program mode, the program mode shall be exited. If at any point the scan button is pressed and held for a period of 10 seconds, the unit shall enter the service mode.