Starting the ETD Module after a Complete Shutdown
Issue
Starting the ETD module after a complete shutdown
Environment
LTQ XL Ion Trap Model
Resolution
To start up the ETD Module after a complete shutdown:
- Start the LTQ Orbitrap XL ETD according to the start up procedures given in “Starting Up the System after a Shutdown“ above. This also turns on the ETD Module as the ETD Module power controls are linked to the MS power controls.
- If the Tune Plus window is not already open, choose Start > Programs > Thermo Instruments> LTQ> LTQ Tune from the taskbar. The Tune plus window will open.
NOTE: You can determine the state of the MS detector by observing the state of the On/Off/Standby button on the Control/Scan Mode toolbar.