Thermo Fisher Scientific
Recommended Windows Settings for Exactive Systems
Issue 
Upgrade to a Windows 10 computer from a Windows 7 computer
Environment 
Windows operating system installed on these instruments:
  • Exploris Series
  • Eactive Plus
  • Exactive Plus EMR
  • Q Exactive
  • Q Exactive Plus
  • Q Exactive HF
  • Q Exactive Focus
  • Q Exactive HF-X
  • Q Exactive UHMR 
Resolution 
All Exactive series instruments (except Exactive classic) can be upgraded from Windows 7 to Windows 10 2019 LTSC. Only Windows 10 2016 LTSB and 2019 LTSC are compatible with these instruments.
For this upgrade, please order the new PC (Dell Optiplex XE3) with Windows 10 2019 LTSC, using the part number BRE0023909. The software is not pre-configured, so please follow the steps mentioned below to correctly configure the new PC. It is not required to have an internet connection for this setup, just the time synchronization will fail, which is fine. However, Internet connection is required for downloading the required software, which can be then transferred to the customer PC via USB or other means. It is divided into 3 parts, please follow them serially.
Recommended Windows Settings
Follow these steps to verify the settings were properly applied. 
This section shows the related Windows settings for the each script item as reference. It is optional to modify these settings. 
Power Options 
Under Control Panel > Hardware and Sound > Power Options the recommended setting for the power plan are listed below. 
Figure 18.jpg
Click Change plan settings to view the settings. 
Figure 19.jpg
Click Change advanced power settings to view the settings. 
Advanced power settings are: 
Figure 20.jpg
USB Power Management 
Under Control Panel > Hardware and Sound > Devices and Printers > Device manager: “Allow the computer to turn of this device to save power” needs to be unchecked for each of the device under Universal serial Bus controller with Power Management Settings. 
Figure 21.jpg
Network Card Power Management 
The boxes under Power Management for the Network card should be un-checked. 
Figure 34.png
Local Group Policy Windows Updates 
For doing so, [Windows Key]+R has to be pressed, and “gpedit.msc” has to be executed. Under Local Computer Policy > Computer Configuration > Administrative Templates > Windows Components > Windows Update, the following settings have to be chosen. Right-click on each setting and choose Edit to open the dialog box. 
Enabling Windows Update Power Management to automatically wake up the system to install scheduled updates: Select Disabled. Click on OK. 
Figure 22.jpg
Configure Automatic Updates: Select Enabled and Configure automatic updating option 3 - Auto download and notify for install. Click OK. 
Figure 23.jpg
No auto-restart with logged on users for scheduled automatic updates installations: Select Enabled. Click OK. 
Figure 24.jpg
Give me updates for other Microsoft products when I update Windows 
Under Settings > Update & Security > Windows Update > Advanced options: Turn On “Give me updates for other Microsoft products when I update Windows” 
Figure 25.jpg
Disable Remote Assistance 
Under Control Panel > System and Security > System > Advanced system settings: uncheck Allow Remote Assistance connections to this computer and click Don’t allow remote connections to this computer. 
Figure 26.jpg
Telnet 
Under Control Panel > Programs > Programs and Features > Turn Windows features on or off: check Telnet Client 
Figure 27.jpg
Network Adapter Internet 
Under Control Panel > Network and Internet > Network Connections: configure and rename the connection for Intel(R) Ethernet Connection I219-LM 
Figure 28.jpg
Network Adapter Instrument 
Under Control Panel > Network and Internet > Network and Sharing Center > change adapter settings: configure and rename the connection for Intel(R) Ethernet Server Adapter I210-T1. 
Figure 29.jpg
.NET Framework 3.5 
Under Control Panel > Programs > Programs and Features > Turn Windows features on or off: check .NET Framework 3.5 (includes .NET 2.0 and 3.0) 
Figure 30.jpg
.Net WCF MSMQ Activation 
Under Control Panel > Programs > Programs and Features > Turn Windows features on or off: check .NET Framework 4.6 Advanced Services > WCF Services > Message Queuing (MSMQ) Activation 
Figure 31.jpg
Rename Computer 
Caution should be used when changing the name of the computer if SII or Chromeleon is already installed.
Under Settings > System > About: match the PC name with the Dell Service Tag. Restart the computer.
Note: New name is listed on this page and will change when the computer is restarted 
Figure 32.jpg
Time Synchronization 
Under Settings > Time & language > Date & time: set the current date and time. To manually Change date and time, turn Off the setting Set time automatically. 
Figure 33.jpg
Homepage of Internet Explorer
Set the homepage of Internet Explorer to https://corporate.thermofisher.com.