European Pharmacopoeia 9.7 2.2.40 Compliance
Issue
Are Thermo Scientific FTIR spectrometers compliant with European Pharmacopoeia (Ph. Eur.) regulations?
Environment
FT-IR Spectrometers
Resolution
As of April 1, 2019, Thermo Scientific FTIR spectrometers are compliant with the new European Pharmacopeia 2019 regulations. Our internal and external standards are built with NIST traceable polystyrene. Our calibration lab tests all standards and reports the location of all 14 NIST traceable peak locations, including those required by Ph. Eur. 9.7 2.2.40.
See the attached customer letter for more details.
Attachment(s)
File | Last Modified |
---|---|
European Pharmacopoeia 9.7 2.2.40 Compliance - Customer Letter.pdf | December 25, 2021 |