Sampling Accessories for the Summit Spectrometer
Nicolet Summit FTIR spectrometers are compatible with Thermo Scientific™ Nicolet™ iS5 iD sampling accessories and hundreds of other third-party accessories.
For instructions on installing a sampling accessory, see Install a Sampling Accessory
With the following accessories and adapters, the Summit spectrometer can quickly and easily measure a wide range of samples.
Everest ATR Accessory
The Everest™ ATR accessory is a high-performance, versatile single-bounce attenuated total reflectance accessory and is ideal for analyzing liquids, solids, pastes, or powders. The Everest ATR accessory was specifically designed to fit Summit spectrometers.
Contact us for more information.
iD1 Transmission Accessory
The iD1 Transmission accessory is convenient for sampling films, pellets, liquids, and gases and fits a wide range of transmission accessories. The sample compartment fits gas cells and cuvettes up to 10 cm.
Contact us for more information.
iD5/iD7 ATR Accessories
The iD5 and iD7 ATR accessories—designed for the Nicolet iS™5 spectrometer—are alternatives to the Everest ATR accessory. While these ATR accessories are excellent multi-purpose accessories, the Everest ATR accessory is recommend for use with Summit spectrometers.
iD Base Adapter Plate
The iD Base Adapter accommodates a wide variety of third-party accessories for FTIR sampling, including large transmission accessories and other specialized accessories. Pair the iD Base Adapter with the slide mount sample holder for even more options.
Slide Mount Sample Holder
Pair a slide mount sample holder with the iD Base adapter to use slide-mounted accessories that are too large for the iD1 Transmission accessory.
Contact us for more information.
Golden Gate ATR Accessory
The Golden Gate ATR accessory features a sapphire anvil for high pressure contact and a rugged exterior, allowing you to analyze a range of samples, from single particles and fibers or hard solids to corrosive liquids. The Golden Gate ATR accessory is also ideal for macro sampling.
Attachment(s):
| File | Last Modified |
|---|---|
| 115360_iD1 Transmission Open Cover with Poly.jpg | July 22, 2022 |
| 115356_Golden Gate ATR.jpg | July 22, 2022 |
| 115358_iD Base and Slide Mount.jpg | July 22, 2022 |
| 115357_iD Base Adapter_1.jpg | July 22, 2022 |
| 115361_iD7 ATR accessory front.jpg | July 22, 2022 |
| 115359_iD1 transmission inside with card.jpg | July 22, 2022 |
| 115355_Everest ATR front white background.jpg | July 22, 2022 |