Mercury TGA or Mercury GC Search Gives Error Message "The series data cannot be used in this function"
To resolve the Mercury TGA or GC error in OMNIC v9, ensure the series data was collected using the correct Data Collection Type—either TGA/IR or GC/IR—based on the intended analysis.
Issue
When trying to start a Mercury TGA or Mercury GC analysis, the following message appears: "The series data cannot be used in this function. Please make sure the correct type of series data is selected."
Environment
OMNIC v9
Resolution
When performing a Mercury TGA or Mercury GC analysis in OMNIC, the series data being used must have been collected with the appropriate Data Collection Type.
To perform the Mercury TGA analysis, make sure the Data Collection Type setting on the Series tab of Experiment Setup is set to "TGA/IR".
To perform the Mercury GC analysis, make sure the Data Collection Type setting on the Series tab of Experiment Setup is set to "GC/IR".
Analysis
With a series data set that has already been collected, the collection type can be verified by:
- Opening the series file
- Clicking on the Series menu and the Show Series Info command
- Checking in the Data Processing History section of the Collection and Processing Info.
Attachment(s)
| File | Last Modified |
|---|---|
| Verify Series Collecton Type.jpg | June 22, 2024 |
| GC-IR Selection.jpg | June 22, 2024 |
| TGA-IR Selection.jpg | June 22, 2024 |
| Mercury TGA or GC Error.JPG | June 22, 2024 |