Thermo Fisher Scientific
QCheck Algorithm
QCheck uses a derivative-based correlation algorithm to compare unknown spectra with library entries, applying a scaling function to enhance sensitivity and report similarity metrics.
Issue
What is the QCheck Algotrithm?
Environment
OMNIC
Resolution
Refer to the attachment for more details.
  • Was this article helpful?