Explore Measurements in Spectra View
When viewing your measurement results in the session view, you may want to explore a specific spectrum in more detail with tools not available in the session view. Fortunately, you can easily save a spectrum to your Measurements and then open it in the Spectra view.
❖ To view a spectrum from your session analysis in the Spectra view
- In the Session Analysis view tab, select the Cursor tool and click a point you wish to analyze.
- Open the spectra pane, if it is not already open, to view the spectrum at the selected point.
- Right-click the spectrum and select Add selected spectrum to dashboard measurements. Note the X and Y position of the spectrum, as displayed in the spectra pane.
- Click the Dashboard icon to return to the dashboard.
- Under Measurements, find your spectrum. The spectrum is named using the X and Y position.
- Right-click the spectrum and open it in the Spectra view.
Export spectra with the split tool
With the Split tool, you can export all spectra from one or more regions. Save the spectra as SPA files for sharing or analysis with other tools, or save them to the dashboard or view them in the spectra tab.
To export spectra with the split tool
-
In a session analysis tab, open the region queue.
-
Select the measurements to split and click Split.
-
Choose to send the exported spectra disk, to the dashboard, or to a tab. Spectra sent to the dashboard or a spectra tab are saved to the database. Spectra sent to disk are saved as SPA files.
Note: The software will ask for confirmation before proceeding, as splitting large area maps can take time and require significant data storage.