Normalize spectra scale
Use Normalize to change the Y-axis scale of the selected spectra to a “normal” scale in which the Y values of the data points range from 0 absorbance units for the lowest point to 1 absorbance unit for the highest peak (for an absorbance-like spectrum) or from 10% to 100% transmittance (for a transmission-like spectrum). These normal scales are typical of spectra in commercial spectral libraries.
After you normalize the scale of a spectrum, you cannot use it for quantitative analysis unless you know the scaling factor and adjust the quantitative results accordingly.
Desktop interface
To normalize the scale in the desktop interface, from a Spectra tab, go to Process > Normalize.
Touchscreen interface
To normalize the scale in the touchscreen interface, from the Spectra view, open the main menu and select Spectral Corrections > Normalize Scale.