Thermo Fisher Scientific
Print Screen with CTRL+P
Users can press CTRL+P to quickly print the current display in any spectra, session, or series tab, with optional adjustments in Page Setup. For detailed control, generating a report is recommended.
Quickly print the current display using the CTRL+P keyboard shortcut. This feature is available on any spectra, series, or session analysis tab.
To print
1. When viewing data in a spectra, session, or series tab, press CTRL+P to bring up the print preview.
2. Use Page Setup to change orientation, margins, and paper size.
In a spectra tab, use the Settings tab to show or hide additional data, which will be included in the printed image when using CTRL+P. For more control over which data is printed, create a report instead of using this keyboard shortcut.
  • Was this article helpful?