Thermo Fisher Scientific
Replacing tubing and fittings
Issue
Quick tips on how to replace tubing and fittings
Environment
Thes ASE 350 models:
  • B51004319
  • 083114
  • 083115
  • 083146
  • 083151
  • 083152
  • 083153
  • 083157
  • 082984
Resolution
Tips on replacing tubing and fittings:
  • Use a 1/4-inch open-end wrench (P/N 014605) to tighten any leaking fittings. Do not overtighten!
  • If tightening a fitting does not stop a leak, the tubing and fitting assembly must be replaced.
  • Was this article helpful?