Thermo Fisher Scientific
Fitting the molecular sieve drying trap
Issue 
How do I fit the molecular sieve drying trap?
Environment
  • iCAP 6500 Radial
  • iCAP 6500 Dual
  • iCAP 6300 Radial
  • iCAP 6200 Dual
  • iCAP 6300 Dual
Resolution
  The molecular sieve dying trap is mounted in the support on the side cover of the RF generator.
  • Secure it in place with 2 ty-raps.
  • Connect the camera purge gas from the gasbox to the inlet at the bottom.
  • Connect the outlet at the top to the camera.
Note – The orientation is not important when the trap is first installed. However, if it is removed for any reason, it must be replaced in the same orientation.