Thermo Fisher Scientific
POP Window Cleaning
Issue
Correction of low intensities by cleaning the POP windows
Environment
  • iCAP 6000
  • iCAP 7000
Resolution
Please, see the attached procedure below.
Cleaning iCAP OES windows.pdf
Analysis
Low intensities on radial or axial view.
Cause
The POP window is dirty allowing minimal light to pass.
  • Was this article helpful?