ASX-560 LED Flashing Red
Issue
Cetac ASX-560 autosampler probe will not move. Power LED is flashing red.
Environment
iCAP Qnova Series
iCAP 7000 Series
iCAP PRO series
Cetac ASX-560
Cetac ASX-280
Resolution
Replace autosampler. If it is an ASX-560 try using P/N EXBRE0003260 before ordering new.
Analysis
Cause
Most likely one of the sensors on the autosampler has failed