Intermittent Loss of Communication with Autosampler
Issue
Intermittent loss of communication with Cetac ASX-560 autosampler
Environment
Cetac ASX-560
Cetac ASX-289
Resolution
Moved USB cable which connects autosampler to the computer from a USB 3.0 port (typically identified by a blue insert within the port connector - see image to follow) to a USB 2.0 port. Please reference iCAP Q Service Alert Pi 112.
USB 3.0 port
Reconfigured COM port settings for ASX-560 to reflect the new COM port the autosampler set up on. This knowledge article can ge referenced to complete this task: Adding Autosampler in Qtegra
Enabled all of the MSMQ features using this knowledge article: Qtegra for ICP-MS and ICP-OES Software Issues
Analysis
Communication would seemingly restore on its own without any user invovlement but due to the loss of communication in the first place the LabBook would become interrupted and therefore the analysis would cease.