Thermo Fisher Scientific
FAST Valve Maintenance
Issue
FAST and Prepfast Valve Maintenance
It is recommended to clean the stator and rotor on the PrepFAST/FAST valve(s) on a weekly basis if it is being used to analyze samples 8 hours a day / 5 days a week and bi-weekly if the frequency of usage is less. This video will show you how to perform maintenance on the valve.
Environment
  • PrepFAST
  • FAST
  • iCAP Q
  • iCAP Qnova
  • iCAP 7000
  • iCAP PRO
Resolution
Analysis
Cause
  • Was this article helpful?