Labbook Freezes while using PrepFAST
Issue
Labbook Freezes on "Wash will complete when the FAST method finishes."
Environment
- iCAP Qnova
- iCAP Q
- ESI PrepFAST SC-4dx
Resolution
- Open the FAST method being used
- Find the line that says "Loop Rinse Toggle"- This will be in the right column of the FAST method
- Click the + button to expand that line
- Change the timing for Method at 12s to 11s, Method at 18s to 12s. (Double click on each line, and a box will appear for you to change these values.)
- Once changed, click save to save the FAST method.
- Once saved, restart the computer, and reload Qtegra. Make a new lab book and reload the FAST method. Double check that the new values are correct.
NOTE: If using a 1.5mL loop you may also want to check the method settings and make sure that the uptake time is between 30 and 35s.
Analysis
Cause
Qtegra update, requires this change.
Attachment(s)
File | Last Modified |
---|---|
clipboard_ed6e24d037ab3564f587d5157962f2d60.png | July 05, 2022 |
clipboard_e171c9e0369b5d401573d32dec63e923a.png | July 05, 2022 |