Manually Adjusting SC Autosampler Speeds in Qtegra
Issue
Speed of axis needs to be adjusted to for example resolve a movement issue being experienced
Environment
- SC2-DX
- SC4-DX
- SC8-DX
- SC16-DX
Resolution
1. Open Configurator
2. Expand the configuration being used for analysis
3. Right-click on ESI plugin and select Edit settings
4. Click Settings and then select Motor
5. Adjust the appropriate motor speed
6. Click Save
Attachment(s)
File | Last Modified |
---|---|
clipboard_e0dfee6cbb0b523ce12094b75f11f7c48.png | January 04, 2024 |