No Flow Through Valve
Issue
No solution is getting to the nebulizer from the FAST valve
Environment
- FAST
- PrepFAST
Resolution
Check to ensure that the valve has been assembled correctly. The P6/P7/P7+ marker should be in line with ESI marking and also in line with the screw head in between ports 4 and 5.
If the valve has been assembled correctly, then it is possible that the actuator is not snapping into the correct position when sent to the Load and Injector position. To correct this try re-indexing (re-homing) the valve. This knowledge article will instruct you on how to re-home the valve: Re-Homing the Valve
Cause
If the valve is not assembled correctly, the ports in the stator will not be aligned properly with the correct groove in the rotor.
Attachment(s)
File | Last Modified |
---|---|
PrepFAST Valves Installed Correctly.png | November 03, 2022 |
P6 Valve.gif | November 03, 2022 |