FAST Valve Maintenance
Issue
FAST and Prepfast Valve Maintenance
It is recommended to clean the stator and rotor on the PrepFAST/FAST valve(s) on a weekly basis if it is being used to analyze samples 8 hours a day / 5 days a week and bi-weekly if the frequency of usage is less. This video will show you how to perform maintenance on the valve.
Environment
- PrepFAST
- FAST
- iCAP Q
- iCAP Qnova
- iCAP 7000
- iCAP PRO
Resolution
Analysis
Cause