Thermo Fisher Scientific
Using AI1310, AS1310 Autosampler in High Throughput Mode
Mount two AS1310 samplers on the GC, each connected separately to the PC. Connect the samplers to each other with a sync cable and to the GC with a Remote Start "Y" cable. Configure the samplers in Chromeleon, creating separate instruments for the front and back, then link them to the TRACE 1300 GC. Ensure proper communication and set the GC to "Shared" mode. Create and synchronize instrument methods and sequences for high-throughput operation.
Issue
How to use AI/AS1310 Autosampler in High Throughput mode
Environment
  • AI 1310
  • AS 1310
  • Chromeleon
Resolution
Step 1: Hardware configuration:
  • Two AS1310 samplers are mounted on the GC, one over each inlet. A special mounting bracket is used.
  • Each sampler is connected to the PC using its own, separate serial cable; do not use the “Y” serial cable in the Gemini kit. Note that this requires either:
  • Two available RS-232 ports on the PC, or
  • One available RS-232 ports on the PC + the on-board serial port on the back of the GC
  • The two samplers should also be connected to each other using the Sync cable
  • Finally, the two samplers should be connected to the GC using the Remote Start “Y” cable.
Figure 1: Connection
HTM figure 01.jpg
Step 2: Software Configuration:
  • Open the Chromeleon Instrument Configuration Manager
  • Create a new instrument, using a name that identifies it as the Front portion of the GC
  • Add an AI/AS 1310 Sampler to this instrument and configure it normally. Be sure to specify the communications port of the sampler mounted on the Front Inlet
  • Do not add a GC at this time
  • Create a second new instrument, using a name that identifies it as the Back portion of the GC
  • Add an AI/AS 1310 Sampler to this instrument and configure it normally. Be sure to specify the communications port of the sampler mounted on the Back Inlet
  • Do not add a GC at this time
  • Ensure the Back Instrument is selected, and add a TRACE 1300 GC.  Specify the IP address and use the ‘Get’ button to ensure a proper configuration
  • In the TRACE 1300 GC Configuration dialog, select the ‘Instrument’ tab and change the Front Instrument to the name you previously specified for the Front Instrument
  • Verify that the signal/hardware handshake settings match the following:
    • Remote start in: HighToLow
    • End of run out: HighToLow
    • Inhibit ready: None
    • GC ready out: Low
    • Start of run out: HighToLow
    • Prep-run out: Low 
  • After pressing OK, your configuration should look similar to the Figure 4.
  • Start Chromeleon
  • Test communications to ensure the configuration is correct:
    • The GCs of both instruments should be connected and reporting status read-backs  (identical values of course)
    • The autosamplers of both instruments should be connected
    • For each GC, press the F8 button. In the dialog that appears, highlight the ‘Sampler’ in the left pane and select the ‘Command’ tab in the right pane.
    • Click the ‘Wash’ command and ensure that the sampler configured for each GC properly washes.
  • Set the GC to Operate in ‘Shared’ Mode:
    • Select the Front GC
    • Press the F8 button. In the dialog that appears, highlight the ‘GC’ in the left pane
    • Find the ‘Dual Operation’ property in the right pane and set the value to ‘Shared’
Figure 2: Instrument Configuration
HTM figure 02.jpg
Figure 3: 'Instrument' Tab
HTM figure 03.jpg
Figure 4: Instrument Configuration
HTM figure 04.jpg
Figure 5: GC Washes
HTM figure 05.jpg
Figure 6: Dual Operation Properties
HTM figure 06.jpg
Step 3: Instrument Method Setup:
Front Instrument Method:
  • Create an instrument method for the Front Instrument in the normal way.
  • When the wizard completes, select the ‘Script Editor’ tab
  • In the {Initial Time} section of the script, right-click and select ‘Insert Command’
  • Choose the ‘GC.Sharable’ command, and set the value to ‘On’.
Back Instrument Method:
  • Create an instrument method for the Back Instrument in the same way as previously done for the Front Instrument.
  • The parameters for the Oven and any other shared components for the two instrument method must be identical.
  • Be sure to insert the ‘GC.Sharable’ command, and set the value to ‘On’.
Figure 7: Instrument Method
HTM figure 07.jpg
Step 4 : Sequence Setup
  • Create two sequences – one for the Front GC, and one for the Back GC  - as you normally would
  • Be sure to use the appropriate instrument method for each sequence
  • Important: Before staring the sequences, confirm that the GC is in ‘Shared’ mode
  • Start the first sequence
  • The sequence state will become ‘Running’, but the GC will remain in Stand-by mode and no injection will occur
  • Start the second sequence
  • The sequence state will become ‘Running’, and both autosamplers will begin their injection cycle when the GC becomes ready.
Limitations
  • Rebooting the PC and/or power-cycling the GC can cause the ‘Dual Operation’ property of the GC to be reset to ‘Front Only’. Always be sure to check the property before submitting Gemini mode sequences
  • Because the two sequences are running on a shared GC, any hardware errors or actions that cause one sequence to abort will result in both sequences being aborted
  • Gemini Mode is fully supported using the Chromeleon “SE for GC” license.
  • Was this article helpful?