TurboFlow Column Wash Solvent
Issue
How do I prepare the TurboFlow Column wash solvent?
Environment
- LX-2
- LX-4
Resolution
Prepare a 45/45/10 acetonitrile/isopropanol/acetone solution in a clean 2-liter bottle by mixing the following:
• 900 mL of LC/MS-grade acetonitrile
• 900 mL of LC/MS-grade isopropanol
• 200 mL of LC/MS-grade acetone
NOTE:
- Use this solution to fill the column wash solvent reservoir.
- The solution is stable for 30 days at room temperature.
- You can use column wash solvent as the autosampler's organic wash solution when carryover is an issue.