Thermo Fisher Scientific
How to Replace the Flow Cell
Issue
Damaged or leaking detector flow cell
Environment
Vanquish HPLC and UHPLC systems
Resolution
If you see that the flow cell of your detector is damaged or leaking, it should be replaced.
Required parts:
  • New flow cell
Preparation steps:
  1. Flush the flow cell with isopropanol
  2. Wait until the flow cell has cooled down
Analysis
The Vanquish HPLC and UHPLC systems offer uncompromised performance, robustness, and ease of use. A key element of its user-friendly functionality is the accessibility of the system for simple maintenance and replacement procedures. These videos provide step-by-step instructions on how to perform these procedures and keep your Vanquish HPLC and UHPLC systems operating smoothly. To access these procedures from your instrument, without the need for an external internet connection, consider adding the Vanquish User Interface to your system.
  • Was this article helpful?