Thermo Fisher Scientific
How to clear rack information
Issue
How do I clear rack information?
Environment
  • Gallery
  • Gallery Plus
  • Beermaster Analyzer 6.0
  • Beermaster Analyzer 7.0
Resolution
To clear the information from all positions from a rack:
NOTE: The rack has to be outside of the analyzer when the rack information is handled. 
  1. Select F3 > REAGENT RACKS. 
  2. Select the rack from the racks field. 
  3. Select the rack position.
  4. Click 'clear position' to remove the rack position information. 
NOTE: The function is useful, for example, when the rack has been removed from the analyzer, but the rack information still exists in the user interface.
  • Was this article helpful?