Thermo Fisher Scientific
Import TGA File
If you are running a TGA experiment with a TA Instruments analyzer, you can import the TGA data into your series collection to add weight, temperature, and derivative weight to the results. Export the data as a .tga file and import it via File > Time Series > Import TGA File.
If you are running a TGA experiment with a TA Instruments analyzer, you can import the TGA data to your series collection to add weight, temperature, and derivative weight to the results.
import tga file.png
To import TGA data
  1. Export the data from the TGA instrument as a file with a .tga extension. See the documentation that came with your analyzer for instructions on how to export the tga data.
  2. While viewing the series data, go to File > Time Series > Import TGA File.
  3. Select the TGA file and select Open.
    The temperature, weight, and derivative weight profiles are added to the Profiles tab.
If you are running a TGA experiment with a TA Instruments analyzer, you can import the TGA data into your series collection to add weight, temperature, and derivative weight to the results. Export the data as a .tga file and import it via File > Time Series > Import TGA File.
  • Was this article helpful?