Rinse Solution is Not Flowing into Rinse Station
Issue
The rinse solution is not flowing into the rinse reservoir.
Environment
- ASX-560
- ASX-280
Resolution
Try increasing the speed of the rinse pump. This can be done as follows:
1) Open Configurator
2) Click on Experiment Configurator
3) Locate the configuration you use and click on triangle to expand it and reveal plugins
4) Right-click on ASX-560 and click Edit settings
5) Increase the Analysis Pump Speed. It defaults to 24 (percentage of maximum speed). You could try 50 for example.
6) Save the configuration
If the increase in pump speed fails to cause the rinse solution to start flowing into the resevoir, you can also try increasing the tension on the rinse pump tubing using this knowledge article as a reference to do so: Adjusting the Tension on the Peristaltic Pump for Cetac ASX-560 and ASX-280 Autosamplers
Cause
The rinse pump is not turning fast enough to draw the solution up from the rinse vessel. This could especially be true if the distance is longer from the rinse vessel to the pump. Also, there may not be enough tension applied to the rinse pump tubing to cause it to make contact with the pump rollers and thus draw up liquid.
Attachment(s)
| File | Last Modified |
|---|---|
| Save configuration.PNG | January 17, 2023 |
| Analysis Pump Speed.PNG | January 17, 2023 |
| Edit Settings.PNG | January 17, 2023 |
| Expand Configuration.PNG | January 17, 2023 |
| Experiment Configurator.PNG | January 17, 2023 |
| Configurator.PNG | January 17, 2023 |