Thermo Fisher Scientific
Experiencing gas and air leaks
Issue
Leak during extraction
Environment
Thes ASE 350 models:
  • B51004319
  • 083114
  • 083115
  • 083146
  • 083151
  • 083152
  • 083153
  • 083157
  • 082984
Resolution
If you suspect a leak, check the following locations:
  1. The rear panel
  2. The solvent reservoirs
  3. The connections inside the service area
Tips:
  • Listen for leaks, and run your hand over connections to feel for escaping gas.
  • If you find a loose connection, push the tubing firmly onto its fitting.
  • If the fitting or tubing continues to leak, replace it.
  • Was this article helpful?