Thermo Fisher Scientific
Correcting Restrictions
Issue
How can I correct restrictions? 
Environment
  • Integrion models:
    • 22153-60301
    • 22153-60302
    • 22153-60303
    • 22153-60304
    • 22153-60305
    • 22153-60306
Resolution
Possible Causes and Solutions:
1. Tubing or fitting
  • Back flush or replace the tubing or fitting. Be careful not to overtighten fittings.
2. Column
  • Replace the column inlet bed support or clean the column.
  • For instructions, refer to the column manual provided on the Thermo Scientific Reference Library DVD (P/N 60-053891).
  • If sample precipitation adsorbing onto the column is suspected, filter or dilute the sample, or run the sample through an OnGuard cartridge or InGuard cartridge.
3. Injection valve 
  • If the injection valve is the suspected source of the high backpressure, observe a run to see whether the high-pressure limit is triggered when injection occurs; if it is, the injection valve rotor seal or stator may need to be replaced.
4. Dionex EGC III 
  • Replace the outlet frit as instructed in the Dionex EGC manual.
  • The manual is provided on the Thermo Scientific Reference Library DVD (P/N 60-053891).