Thermo Fisher Scientific
System showing zero ion counts
Issue
IC System showing zero Ion counts, (0%) on a new EG cartridge.  
Environment
All of the reagent-free Ion chromatography systems
Resolution
  1. Find the latest firmware and update it from the service manager in the software apply a a concentration value on the EG and monitor the current-voltage value in the software command, check the change in the current value when you change the concentration applied values. 
  2. If this still showing zero value, EG control board need to be checked and replaced.
Cause
  • System not recognizing the EG cartridge because of its firmware
  • Faulty EG control board/power supply
  • Was this article helpful?